Assay Design
Gene Transcripts
- Gene Symbol
- Triml1
- Entrez Gene ID
-
290767
- Gene Name
- tripartite motif family-like 1
- Gene Aliases
-
RGD1305337
- Chromosome Location
-
Chr.16: 52167989 - 52177522 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_224885.7 |
XP_224885.3 |
3 - 4 |
1003 |
82 |
View Full Product Details