Assay Design



Gene Transcripts

Gene Symbol
Csdc2
Entrez Gene ID
266600
Gene Name
cold shock domain containing C2, RNA binding
Gene Aliases
Pippin
Chromosome Location
Chr.7: 123168811 - 123183336 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170542.1 NP_001164013.1 2 - 3 504 73
GenBank mRNA CO405729.1 - 2 - 3 504 73
X89962.1 - 2 - 3 403 73

View Full Product Details