Assay Design



Gene Transcripts

Gene Symbol
Invs
Entrez Gene ID
313228
Gene Name
inversin
Gene Aliases
RGD1563359
Chromosome Location
Chr.5: 64031131 - 64180830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107932.1 NP_001101402.1 13 - 14 2943 73

View Full Product Details