Assay Design



Gene Transcripts

Gene Symbol
Pf4
Entrez Gene ID
360918
Gene Name
platelet factor 4
Gene Aliases
Cxcl4, PF-4, Pf4a, RATPF4A
Chromosome Location
Chr.14: 18848549 - 18849258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007729.1 NP_001007730.1 2 - 3 230 77

View Full Product Details