Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199384.2 | NP_955416.1 | 1 - 2 | 195 | 72 |
GenBank mRNA | BC063179.1 | - | 1 - 2 | 199 | 72 |
FQ212732.1 | - | 1 - 2 | 597 | 72 | |
FQ213363.1 | - | 1 - 2 | 229 | 72 | |
FQ232171.1 | - | 1 - 2 | 253 | 72 | |
FQ232930.1 | - | 1 - 2 | 213 | 72 | |
FQ235113.1 | - | 1 - 2 | 100 | 72 |