Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199384.2 | NP_955416.1 | 6 - 7 | 707 | 72 |
GenBank mRNA | BC063179.1 | - | 6 - 7 | 711 | 72 |
FQ212732.1 | - | 6 - 7 | 1109 | 72 | |
FQ213363.1 | - | 6 - 7 | 741 | 72 | |
FQ232930.1 | - | 6 - 7 | 725 | 72 | |
FQ235113.1 | - | 6 - 7 | 612 | 72 |