Assay Design



Gene Transcripts

Gene Symbol
Morn2
Entrez Gene ID
500606
Gene Name
MORN repeat containing 2
Gene Aliases
RGD1563988
Chromosome Location
Chr.6: 2968297 - 2974419 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126086.2 NP_001119558.1 1 - 2 92 89
GenBank mRNA BC166893.1 - 1 - 2 71 89
FM039224.1 - 1 - 2 55 89

View Full Product Details