Assay Design



Gene Transcripts

Gene Symbol
Trmt5
Entrez Gene ID
362754
Gene Name
tRNA methyltransferase 5
Gene Aliases
RGD1306567
Chromosome Location
Chr.6: 96164579 - 96171678 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108713.1 NP_001102183.1 4 - 5 888 94
XM_006240213.2 XP_006240275.1 3 - 4 864 94

View Full Product Details