Assay Design



Gene Transcripts

Gene Symbol
Pqlc3
Entrez Gene ID
298906
Gene Name
PQ loop repeat containing 3
Gene Aliases
RGD1306818
Chromosome Location
Chr.6: 42290678 - 42302577 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034952.1 NP_001030124.1 6 - 7 684 118
GenBank mRNA CK477351.1 - 6 - 7 507 118

View Full Product Details