Assay Design



Gene Transcripts

Gene Symbol
Wdr72
Entrez Gene ID
300834
Gene Name
WD repeat domain 72
Gene Aliases
RGD1563550
Chromosome Location
Chr.8: 80965734 - 81125710 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135719.1 NP_001129191.1 12 - 13 1571 79

View Full Product Details