Assay Design



Gene Transcripts

Gene Symbol
Ube2u
Entrez Gene ID
500511
Gene Name
ubiquitin-conjugating enzyme E2U (putative)
Gene Aliases
RGD1565480
Chromosome Location
Chr.5: 118574662 - 118642810 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238443.2 XP_006238505.1 3 - 4 614 89

View Full Product Details