Assay Design



Gene Transcripts

Gene Symbol
Serpinb6e
Entrez Gene ID
291087
Gene Name
serine (or cysteine) peptidase inhibitor, clade B, member 6e
Gene Aliases
RGD1561697
Chromosome Location
Chr.17: 32412713 - 32422011 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253875.1 XP_006253937.1 3 - 4 477 130

View Full Product Details