Assay Design



Gene Transcripts

Gene Symbol
Vom2r15
Entrez Gene ID
690680
Gene Name
vomeronasal 2 receptor, 15
Gene Aliases
-
Chromosome Location
Chr.1: 62665651 - 62681471 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099490.1 NP_001092960.1 - 1530 142

View Full Product Details