Assay Design



Gene Transcripts

Gene Symbol
Grap
Entrez Gene ID
363616
Gene Name
GRB2-related adaptor protein
Gene Aliases
-
Chromosome Location
Chr.10: 47930633 - 47949774 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025749.1 NP_001020920.1 4 - 5 530 67
GenBank mRNA BC098740.1 - 4 - 5 530 67
FQ228943.1 - 4 - 5 497 67

View Full Product Details