Assay Design
Gene Transcripts
- Gene Symbol
- Tcrb
- Entrez Gene ID
-
24820
- Gene Name
- T-cell receptor beta chain
- Gene Aliases
-
RATTCB, RATTCBC1, TCB, TCBC1
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC088211.1 |
- |
- |
141 |
150 |
View Full Product Details