Assay Design
Gene Transcripts
- Gene Symbol
- Mup4
- Entrez Gene ID
-
362527
- Gene Name
- major urinary protein 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 77313449 - 77316803 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198784.1 |
NP_942079.1 |
- |
347 |
148 |
GenBank mRNA |
AB039829.1 |
- |
- |
347 |
148 |
View Full Product Details