Assay Design



Gene Transcripts

Gene Symbol
Hs3st3a1
Entrez Gene ID
363618
Gene Name
heparan sulfate (glucosamine) 3-O-sulfotransferase 3A1
Gene Aliases
Hs3st3a, RGD1561262
Chromosome Location
Chr.10: 50928309 - 51014562 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172067.1 NP_001165538.1 1 - 2 561 153

View Full Product Details