Assay Design
Gene Transcripts
- Gene Symbol
- Gabrg1
- Entrez Gene ID
-
140674
- Gene Name
- gamma-aminobutyric acid (GABA) A receptor, gamma 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 39964588 - 40049354 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080586.1 |
NP_542153.1 |
3 - 4 |
422 |
148 |
GenBank mRNA |
X57514.1 |
- |
3 - 4 |
422 |
148 |
View Full Product Details