Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153314.2 | NP_695226.2 | - | 553 | 139 |
XM_006250800.2 | XP_006250862.1 | 1 - 1 | 1647 | 139 | |
XM_008769998.1 | XP_008768220.1 | 1 - 1 | 1653 | 139 | |
GenBank mRNA | M31109.1 | - | - | 553 | 139 |
Y00156.1 | - | 1 - 1 | 597 | 139 |