Assay Design



Gene Transcripts

Gene Symbol
Exo1
Entrez Gene ID
305000
Gene Name
exonuclease 1
Gene Aliases
-
Chromosome Location
Chr.13: 93936989 - 93962749 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107198.1 NP_001100668.1 15 - 15 4856 148

View Full Product Details