Assay Design



Gene Transcripts

Gene Symbol
Alkbh2
Entrez Gene ID
304578
Gene Name
alkB, alkylation repair homolog 2 (E. coli)
Gene Aliases
RGD1306377
Chromosome Location
Chr.12: 48257584 - 48262244 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126273.1 NP_001119745.1 2 - 2 795 112
XM_006249498.2 XP_006249560.1 3 - 3 534 112
GenBank mRNA BC166547.1 - 2 - 2 795 112

View Full Product Details