Assay Design



Gene Transcripts

Gene Symbol
Fastkd2
Entrez Gene ID
301463
Gene Name
FAST kinase domains 2
Gene Aliases
RGD1307883
Chromosome Location
Chr.9: 70448386 - 70471420 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009673.1 NP_001009673.1 7 - 8 1613 141
XM_006245050.1 XP_006245112.1 8 - 9 1770 141
GenBank mRNA BC088416.1 - 7 - 8 1613 141

View Full Product Details