Assay Design
Gene Transcripts
- Gene Symbol
- Crygc
- Entrez Gene ID
-
24277
- Gene Name
- crystallin, gamma C
- Gene Aliases
-
Cryg, Cryg3, Len
- Chromosome Location
-
Chr.9: 71786246 - 71788281 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BG371711.1 |
- |
1 - 1 |
|
97 |
View Full Product Details