Assay Design



Gene Transcripts

Gene Symbol
Rdh13
Entrez Gene ID
361504
Gene Name
retinol dehydrogenase 13 (all-trans/9-cis)
Gene Aliases
-
Chromosome Location
Chr.1: 72956017 - 72983811 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108468.1 NP_001101938.1 7 - 7 2566 103

View Full Product Details