Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107240.1 | NP_001100710.1 | 3 - 3 | 2804 | 164 |
XM_006251554.2 | XP_006251616.1 | 2 - 2 | 2603 | 164 | |
XM_006251555.1 | XP_006251617.1 | 2 - 2 | 2487 | 164 | |
XM_006251556.2 | XP_006251618.1 | 2 - 2 | 2486 | 164 | |
XM_008770427.1 | XP_008768649.1 | 2 - 2 | 4322 | 164 |