Assay Design



Gene Transcripts

Gene Symbol
Tnfrsf11a
Entrez Gene ID
498206
Gene Name
tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Aliases
RANK, RGD1563614
Chromosome Location
Chr.13: 25778306 - 25835802 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271235.1 NP_001258164.1 10 - 10 2420 109
XM_008769487.1 XP_008767709.1 10 - 10 2398 109

View Full Product Details