Assay Design



Gene Transcripts

Gene Symbol
Prm1
Entrez Gene ID
24685
Gene Name
protamine 1
Gene Aliases
-
Chromosome Location
Chr.10: 4945995 - 4946241 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002850.1 NP_001002850.1 1 - 2 106 98
GenBank mRNA BC126070.1 - 1 - 2 190 98

View Full Product Details