Assay Design



Gene Transcripts

Gene Symbol
Krt12
Entrez Gene ID
360625
Gene Name
keratin 12, type I
Gene Aliases
Ka12, Krt1-12
Chromosome Location
Chr.10: 87328547 - 87336709 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008761.1 NP_001008761.1 4 - 5 855 145

View Full Product Details