Assay Design



Gene Transcripts

Gene Symbol
Uba5
Entrez Gene ID
300968
Gene Name
ubiquitin-like modifier activating enzyme 5
Gene Aliases
Ube1dc1
Chromosome Location
Chr.8: 112579455 - 112594192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009669.1 NP_001009669.1 5 - 6 618 145
GenBank mRNA BC088757.1 - 5 - 6 618 145

View Full Product Details