Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024374.4 | NP_077350.1 | 1 - 2 | 183 | 127 |
GenBank mRNA | AY951952.1 | - | - | 190 | 127 |
BC088136.1 | - | 1 - 2 | 183 | 127 | |
D26179.1 | - | 1 - 2 | 238 | 127 | |
FQ228349.1 | - | - | 304 | 127 | |
U21661.1 | - | 1 - 2 | 262 | 127 |