Assay Design



Gene Transcripts

Gene Symbol
Serpinb3
Entrez Gene ID
304688
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 3
Gene Aliases
-
Chromosome Location
Chr.13: 27186701 - 27193210 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008887.1 NP_001008887.1 3 - 4 415 79
XM_006249631.2 XP_006249693.1 5 - 6 637 79

View Full Product Details