Assay Design



Gene Transcripts

Gene Symbol
Vom1r96
Entrez Gene ID
494275
Gene Name
vomeronasal 1 receptor 96
Gene Aliases
V1rb14
Chromosome Location
Chr.4: 121870699 - 121871658 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008927.1 NP_001008927.1 1 - 1 912 84
GenBank mRNA AY510296.1 - 1 - 1 912 84

View Full Product Details