Assay Design



Gene Transcripts

Gene Symbol
Ly49s6
Entrez Gene ID
494196
Gene Name
Ly49 stimulatory receptor 6
Gene Aliases
-
Chromosome Location
Chr.4: 164429048 - 164453171 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009488.1 NP_001009488.1 4 - 5 454 155
GenBank mRNA AY649837.1 - 4 - 5 454 155

View Full Product Details