Assay Design



Gene Transcripts

Gene Symbol
Klra2
Entrez Gene ID
494193
Gene Name
killer cell lectin-like receptor, subfamily A, member 2
Gene Aliases
Ly49i6
Chromosome Location
Chr.4: 165005722 - 165026566 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009718.1 NP_001009718.1 4 - 5 541 131
XM_008763403.1 XP_008761625.1 3 - 4 603 131
GenBank mRNA AY651018.1 - 4 - 5 541 131

View Full Product Details