Assay Design



Gene Transcripts

Gene Symbol
Plscr4
Entrez Gene ID
300900
Gene Name
phospholipid scramblase 4
Gene Aliases
Pls4
Chromosome Location
Chr.8: 99880073 - 99917733 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012000.1 NP_001012000.1 4 - 5 688 134
GenBank mRNA AY548832.1 - 4 - 5 688 134

View Full Product Details