Assay Design



Gene Transcripts

Gene Symbol
Tspyl4
Entrez Gene ID
309828
Gene Name
TSPY-like 4
Gene Aliases
-
Chromosome Location
Chr.20: 41100071 - 41102067 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012075.1 NP_001012075.1 1 - 1 1769 142
GenBank mRNA BC082023.1 - 1 - 1 1769 142

View Full Product Details