Assay Design



Gene Transcripts

Gene Symbol
Krt73
Entrez Gene ID
300248
Gene Name
keratin 73, type II
Gene Aliases
Kb36
Chromosome Location
Chr.7: 143399878 - 143408316 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008808.1 NP_001008808.1 5 - 6 983 179
XM_008765703.1 XP_008763925.1 5 - 6 1023 179

View Full Product Details