Assay Design



Gene Transcripts

Gene Symbol
Ly49i9
Entrez Gene ID
494205
Gene Name
Ly49 inhibitory receptor 9
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009496.1 NP_001009496.1 6 - 6 759 161
GenBank mRNA AY649839.1 - 6 - 6 759 161

View Full Product Details