Assay Design



Gene Transcripts

Gene Symbol
Abcg3l2
Entrez Gene ID
360910
Gene Name
ATP-binding cassette, subfamily G (WHITE), member 3-like 2
Gene Aliases
-
Chromosome Location
Chr.14: 5568023 - 5609333 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014133.1 NP_001014155.1 - 1648 117
GenBank mRNA BC085773.1 - - 1648 117

View Full Product Details