Assay Design



Gene Transcripts

Gene Symbol
Pcdhb5
Entrez Gene ID
291654
Gene Name
protocadherin beta 5
Gene Aliases
-
Chromosome Location
Chr.18: 30398113 - 30400695 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114602.1 NP_001108074.1 1 - 1 1349 148

View Full Product Details