Assay Design
Gene Transcripts
- Gene Symbol
- Ddx19a
- Entrez Gene ID
-
292022
- Gene Name
- DEAD (Asp-Glu-Ala-Asp) box polypeptide 19a
- Gene Aliases
-
Ddx19, ZD10B
- Chromosome Location
-
Chr.19: 43251510 - 43271866 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY303540.1 |
- |
- |
473 |
113 |
View Full Product Details