Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053981.2 | NP_446433.2 | 2 - 2 | 2194 | 122 |
XM_008767763.1 | XP_008765985.1 | 2 - 2 | 9484 | 122 | |
GenBank mRNA | BC127487.1 | - | 2 - 2 | 2194 | 122 |
X78461.1 | - | 2 - 2 | 1918 | 120 |