Assay Design



Gene Transcripts

Gene Symbol
Ka11
Entrez Gene ID
450226
Gene Name
type I keratin KA11
Gene Aliases
-
Chromosome Location
Chr.10: 88141667 - 88144624 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008750.1 NP_001008750.1 4 - 4 761 71

View Full Product Details