Assay Design



Gene Transcripts

Gene Symbol
Wfdc15b
Entrez Gene ID
408230
Gene Name
WAP four-disulfide core domain 15B
Gene Aliases
Pi3, Serpina1c, Skalp, Wfdc14
Chromosome Location
Chr.3: 160607059 - 160617189 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009179.1 NP_001009179.1 2 - 2 120 61
XM_006235606.2 XP_006235668.1 4 - 4 931 61
XM_006235607.2 XP_006235669.1 3 - 3 1196 61

View Full Product Details