Assay Design



Gene Transcripts

Gene Symbol
Kpna5
Entrez Gene ID
294392
Gene Name
karyopherin alpha 5 (importin alpha 6)
Gene Aliases
Ipoa6
Chromosome Location
Chr.20: 32509573 - 32573591 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025113.1 NP_001020284.1 7 - 8 754 102
XM_006256442.2 XP_006256504.1 8 - 9 934 102
XM_008772879.1 XP_008771101.1 6 - 7 609 102
GenBank mRNA AY779029.1 - 7 - 8 754 102

View Full Product Details