Assay Design



Gene Transcripts

Gene Symbol
Tax1bp3
Entrez Gene ID
360564
Gene Name
Tax1 (human T-cell leukemia virus type I) binding protein 3
Gene Aliases
-
Chromosome Location
Chr.10: 59743356 - 59748063 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025419.1 NP_001020590.1 2 - 3 200 89
GenBank mRNA BC097976.1 - 2 - 3 200 89

View Full Product Details