Assay Design



Gene Transcripts

Gene Symbol
Sprn
Entrez Gene ID
541462
Gene Name
shadow of prion protein homolog (zebrafish)
Gene Aliases
-
Chromosome Location
Chr.1: 212619817 - 212623681 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031845.1 NP_001027015.1 1 - 1 163 58
XM_006230573.2 XP_006230635.1 2 - 2 440 58

View Full Product Details