Assay Design



Gene Transcripts

Gene Symbol
Epn2
Entrez Gene ID
60443
Gene Name
epsin 2
Gene Aliases
-
Chromosome Location
Chr.10: 47795496 - 47857338 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033914.1 NP_001029086.1 3 - 4 1024 66
XM_006246521.2 XP_006246583.1 2 - 3 772 66
XM_006246522.2 XP_006246584.1 4 - 5 1106 66
GenBank mRNA BC094524.1 - 3 - 4 1003 66
FQ211574.1 - 4 - 5 1106 66
FQ211939.1 - 3 - 4 1048 66

View Full Product Details