Assay Design



Gene Transcripts

Gene Symbol
Ccdc60
Entrez Gene ID
498190
Gene Name
coiled-coil domain containing 60
Gene Aliases
-
Chromosome Location
Chr.12: 46042422 - 46206748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034945.1 NP_001030117.1 3 - 4 753 67
XM_006249464.2 XP_006249526.1 3 - 4 770 67
GenBank mRNA BC103639.1 - 3 - 4 753 67

View Full Product Details