Assay Design



Gene Transcripts

Gene Symbol
Defb5
Entrez Gene ID
641624
Gene Name
defensin beta 5
Gene Aliases
-
Chromosome Location
Chr.16: 75589530 - 75599524 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037549.1 NP_001032638.1 2 - 2 132 106
GenBank mRNA AY621340.1 - 2 - 2 132 106

View Full Product Details